Organic reference standard, 2,2',3,4,4',5,5'-Heptachlorobiphenyl (PCB No. 180) 10 µg/ml in isooctane, Dr Ehrenstorfer
Catalog # EHERL20018000IO
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 mL
- CAS No.:35065-29-3
- Description:2,2',3,4,4',5,5'-Heptachlorobiphenyl (PCB No. 180) 10 µg/ml in isooctane
- Formula:C₁₂H₃Cl₇
- Description:2,2',3,4,4',5,5'-Heptachlorobiphenyl (PCB No. 180)