Order Entry
Austria
ContactUsLinkComponent
Organic reference standard, 2,2',3,4,4',5,5'-Heptachlorobiphenyl (PCB No. 180) 100 µg/ml in isooctane, CIL
Organic reference standard, 2,2',3,4,4',5,5'-Heptachlorobiphenyl (PCB No. 180) 100 µg/ml in isooctane, CIL
Catalog # PROOCIL-PCB-180-CS
CAS Number:  
Organic reference standard, 2,2',3,4,4',5,5'-Heptachlorobiphenyl (PCB No. 180) 100 µg/ml in isooctane, CIL
Catalog # PROOCIL-PCB-180-CS
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1,2 mL
  • CAS No.:
    35065-29-3
  • Description:
    2,2',3,4,4',5,5'-Heptachlorobiphenyl (PCB No. 180) 100 µg/ml in isooctane
  • Formula:
    C₁₂H₃Cl₇
  • Description:
    2,2',3,4,4',5,5'-Heptachlorobiphenyl (PCB No. 180)

Specifications

About this item