Order Entry
Austria
ContactUsLinkComponent
Organic reference standard, 2,2',3,3',4,5,6'-Heptachlorobiphenyl (PCB No. 174) 35 µg/ml in isooctane, CIL
Organic reference standard, 2,2',3,3',4,5,6'-Heptachlorobiphenyl (PCB No. 174) 35 µg/ml in isooctane, CIL
Catalog # PROOCIL-PCB-174
CAS Number:  
Organic reference standard, 2,2',3,3',4,5,6'-Heptachlorobiphenyl (PCB No. 174) 35 µg/ml in isooctane, CIL
Catalog # PROOCIL-PCB-174
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 mL
  • CAS No.:
    38411-25-5
  • Description:
    2,2',3,3',4,5,6'-Heptachlorobiphenyl (PCB No. 174) 35 µg/ml in isooctane
  • Formula:
    C₁₂H₃Cl₇
  • Description:
    2,2',3,3',4,5,6'-Heptachlorobiphenyl (PCB No. 174)

Specifications

About this item