Order Entry
Austria
ContactUsLinkComponent
 
Black IHC chromogen (HRP), HIGHDEF®
Catalog # ENZOADI9501710030
Supplier:  Enzo Life Sciences
CAS Number:  
Black IHC chromogen (HRP), HIGHDEF®
Catalog # ENZOADI9501710030
Supplier:  Enzo Life Sciences
CAS Number:  

Specifications

  • Pk:
    30 mL
  • Description:
    Black IHC chromogen (HRP)
  • Seq:
    02
  • Storage conditions:
    Long term
  • Storage temperature:
    4 °C

Specifications

About this item

HIGHDEF® black IHC chromogen (HRP) is intended to be used in conjunction with horseradish peroxidase (HRP)-based immunostaining or in situ hybridisation systems.

  • Unique high definition black IHC chromogen for HRP-activated development
  • System used in any automation system as well as manual use
  • Produces a sharply contrasting ebony black colour
Delivery: Black IHC chromogen (HRP) substrate buffer and chromogen in an empty mixing bottle.