Blue IHC chromogen (HRP), HIGHDEF®
Catalog # ENZOADI9501510030
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Pk:30 mL
- Description:Blue IHC chromogen (HRP)
- Seq:02
- Storage conditions:Long term
- Storage temperature:4 °C
Specifications
About this item
Blue IHC chromogen (HRP) is intended to be used in conjunction with horseradish peroxidase (HRP)-based immunostaining or in situ hybridisation systems.
- Unique high definition blue IHC chromogen for HRP-activated development
- Produces a sharply contrasting billiant blue colour
- Easily be distinguished from other stains
Delivery: Substrate buffer and chromogen in an empty mixing bottle.