L(+)-Glutamine for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:56-85-9
:
Formula:C₅H₁₀N₂O₃
MDL Number:MFCD00008044
:146.15 g/mol
Merck Index:13,04484
Boiling Pt:355 °C (1013 hPa)
Melting Pt:185…186 °C
Density:1.5326 g/cm³ (130 °C)
:M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:200-292-1
Specification Test Results
- Solubility:100 g/l (40 °C)
- Melting Point:185 - 186 °C
- Molar Mass:146.15 g/mol
- Bulk Density:640 kg/m3
- pH:4.0 - 6.0 (25 g/l, H2O, 20 °C)
Specification Test Results
Frequently Bought Together