L(+)-Glutamine ≥99%, white powder cell culture reagent
Supplier: MP Biomedicals
Certificates
About this item
CAS Number:56-85-9
:
Formula:C₅H₁₀N₂O₃
MDL Number:MFCD00008044
:146.15 g/mol
Merck Index:13,04484
Boiling Pt:355 °C (1013 hPa)
Melting Pt:185 °C (lit.)
Density:1.5326 g/cm³ (130 °C)
Storage Temperature:Ambient
EINECS:200-292-1
Specification Test Results
- Identity Test:Passes
- Purity:≥99%
- Residue on Ignition:<0.1%
- Loss on Drying:<0.5%
- Optical Rotation:+30 to +35° (c=3.5,1N HCl)
- pH (2%, aq soln):4.5-6.5
Specification Test Results
Frequently Bought Together