Organic reference standard, 2,2',3,4,5,5',6-Heptachlorobiphenyl (PCB No. 185) 10 µg/ml in isooctane, Dr Ehrenstorfer
EHERL20018500IO
:
- Pack type:Glass bottle
- Pk:10 mL
- CAS No.:52712-05-7
- Description:2,2',3,4,5,5',6-Heptachlorobiphenyl (PCB No. 185) 10 µg/ml in isooctane
- Formula:C₁₂H₃Cl₇
- Description:2,2',3,4,5,5',6-Heptachlorobiphenyl (PCB No. 185)