L(+)-Glutamine 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:56-85-9
:
Formula:C₅H₁₀N₂O₃
MDL Number:MFCD00008044
:146.15 g/mol
Merck Index:14,4471
Boiling Pt:355 °C (1013 hPa)
Melting Pt:185 °C (dec.)
Density:1.5326 g/cm³ (130 °C)
Storage Temperature:Ambient
EINECS:200-292-1
Related Information
Frequently Bought Together