Order Entry
Austria
ContactUsLinkComponent
L(+)-Asparagine, anhydrous ≥99.0%, high purity
L(+)-Asparagine, anhydrous ≥99.0%, high purity
Catalog # 94341-100G
Supplier:  VWR Chemicals
CAS Number:  
undefined
L(+)-Asparagine, anhydrous ≥99.0%, high purity
Catalog # 94341-100G
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    100 g
  • Seq:
    3401

Specifications

Specification Test Results

  • Chloride (%):
    <0.05
  • Moisture (%):
    0.5
  • Purity (dried basis) (%):
    99.0
  • Sulfate (%):
    <0.05

Specification Test Results

About this item

 :
 :132.12 g/mol
Merck Index:12,00872
Boiling Pt:343 °C (1013 hPa)
Melting Pt:234…235 °C
Storage Temperature:Ambient
EINECS:200-735-9
CAS Number:70-47-3
Formula:H₂NCOCH₂CH(NH₂)CO₂H
MDL Number:MFCD00064401