Order Entry
Austria
ContactUsLinkComponent
Organic reference standard, 1,4-Difluorobenzene 2000 µg/ml in methanol, Dr Ehrenstorfer
Organic reference standard, 1,4-Difluorobenzene 2000 µg/ml in methanol, Dr Ehrenstorfer
  EHERYA12632000ME
 :  
Organic reference standard, 1,4-Difluorobenzene 2000 µg/ml in methanol, Dr Ehrenstorfer
  EHERYA12632000ME
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    1 mL
  • CAS No.:
    540-36-3
  • Description:
    1,4-Difluorobenzene 2,000 µg/ml in methanol
  • Formula:
    C₆H₄F₂
  • Description:
    1,4-Difluorobenzene