Order Entry
Austria
ContactUsLinkComponent
Pesticide reference standard, 1,2-Dichlorobenzene 1000 µg/ml in methanol
Pesticide reference standard, 1,2-Dichlorobenzene 1000 µg/ml in methanol
Supplier:  Restek
Certificates
Pesticide reference standard, 1,2-Dichlorobenzene 1000 µg/ml in methanol
Supplier:  Restek

Specifications

  • CAS No.:
    95-50-1
  • Formula:
    C₆H₄Cl₂
  • Description:
    1,2-Dichlorobenzene

Specifications

Frequently Bought Together