Order Entry
Austria
ContactUsLinkComponent
 
Organic reference standard, Bromobenzene 2000 µg/ml in methanol
 :  Restek
 
Organic reference standard, Bromobenzene 2000 µg/ml in methanol
 :  Restek

 

  • CAS No.:
    108-86-1
  • Formula:
    C₆H₅Br
  • Description:
    Bromobenzene

 

Frequently Bought Together