Order Entry
Austria
ContactUsLinkComponent
Organic reference standard, Tetrachloroethylene 2000 µg/ml in methanol
Organic reference standard, Tetrachloroethylene 2000 µg/ml in methanol
Supplier:  Restek
Certificates
Organic reference standard, Tetrachloroethylene 2000 µg/ml in methanol
Supplier:  Restek

Specifications

  • CAS No.:
    127-18-4
  • Formula:
    C₂Cl₄
  • Description:
    Tetrachloroethylene

Specifications

Frequently Bought Together