Order Entry
Austria
ContactUsLinkComponent
Blue IHC chromogen (AP), HIGHDEF®
Blue IHC chromogen (AP), HIGHDEF®
Catalog # ENZOADI9501500030
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Blue IHC chromogen (AP), HIGHDEF®
Catalog # ENZOADI9501500030
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    30 mL
  • Description:
    Blue IHC chromogen (AP)
  • Seq:
    02
  • Storage conditions:
    Long term
  • Storage temperature:
    4 °C

Specifications

About this item

Blue IHC chromogen (AP) is intended to be used in conjunction with alkaline phosphatase (AP)-based immunostaining or in situ hybridisation systems.

  • Brilliant blue colour for AP-activated development
  • Formulated for increased stability of the working solution
  • Chromogen substrate system may be used for both automation and manual use
Delivery: Substrate buffer and chromogen in an empty mixing bottle.