Herbicide reference standard, Hexachloro-1,3-butadiene 1000 µg/ml in methanol
Supplier: Restek
Certificates
About this item
Specifications
- CAS No.:87-68-3
- Formula:C₄Cl₆
- Description:Hexachloro-1,3-butadiene
Specifications
Frequently Bought Together