
Iron(II) sulphate heptahydrate
Supplier: Roth Carl
Certificates
About this item
CAS Number:7782-63-0
:
Formula:FeSO₄·7H₂O
MDL Number:MFCD00149719
:278.02 g/mol
Merck Index:13,04091
Melting Pt:64 °C
Density:1.9 g/cm³ (20 °C)
EINECS:231-753-5
:GHS07
Storage Temperature:Ambient
Frequently Bought Together