Order Entry
Austria
ContactUsLinkComponent
Iron(II) sulphate heptahydrate, crystallised, EMPROVE® ESSENTIAL Ph. Eur., BP, USP, FCC, SAFC®
Irritant
Iron(II) sulphate heptahydrate, crystallised, EMPROVE® ESSENTIAL Ph. Eur., BP, USP, FCC, SAFC®
Certificates
Irritant
Iron(II) sulphate heptahydrate, crystallised, EMPROVE® ESSENTIAL Ph. Eur., BP, USP, FCC, SAFC®
Certificates

Specification Test Results

  • Assay (manganometric):
    99.5 - 102.0 %
  • Identity:
    passes test
  • Appearance of solution:
    passes test
  • pH-value (5 %; water):
    3.0 - 4.0
  • Chloride (Cl):
    ≤ 0.01 %
  • Al (Aluminium):
    ≤ 0.01 %
  • As (Arsenic):
    ≤ 0.0002 %
  • Co (Cobalt) (*):
    ≤ 0.002 %
  • Cr (Chromium):
    ≤ 0.0050 %
  • Cu (Copper):
    ≤ 0.0050 %
  • Fe III (Iron III):
    ≤ 0.3 %
  • Hg (Mercury):
    ≤ 0.0001 %
  • Mn (Manganese):
    ≤ 0.1 %
  • Ni (Nickel) (*):
    ≤ 0.0050 %
  • Pb (Lead):
    ≤ 0.0002 %
  • Zn (Zinc):
    ≤ 0.0025 %
  • Residual solvents (ICH Q3C):
    excluded by manufacturing process

Specification Test Results

Related Information

Frequently Bought Together