Order Entry
Austria
ContactUsLinkComponent
 
 
Iron(II) sulphate heptahydrate, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
  0126.0500
 :  Avantor
 :  
 
Iron(II) sulphate heptahydrate, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
  0126.0500
 :  Avantor
 :  

 

  • Pack type:
    Plastic bottle for solids
  • Pk:
    500 g
  • Seq:
    3645

 

 

  • Assay (by KMnO4 titrn.):
    min. 99.0 %
  • Calcium (Ca):
    max. 0.005 %
  • Chloride (Cl):
    max. 0.001 %
  • Copper (Cu):
    max. 0.005 %
  • Potassium (K):
    max. 0.002 %
  • Magnesium (Mg):
    max. 0.002 %
  • Manganese (Mn):
    max. 0.05 %
  • Sodium (Na):
    max. 0.02 %
  • Zinc (Zn):
    max. 0.005 %
  • Ferric ion (Fe³+):
    max. 0.1 %
  • Insoluble Matter:
    max. 0.01 %
  • Phosphate (PO4):
    max. 0.001 %

 

 

 :
 :278.02 g/mol
 :13,04091
 :64 °C
 :1.9 g/cm³ (20 °C)
 :231-753-5
 :GHS07
 :Ambient
 :7782-63-0
 :FeSO₄·7H₂O
 :MFCD00149719