Order Entry
Austria
ContactUsLinkComponent
 
 
Iron(II) sulphate heptahydrate ≥99% ACS
  0387-500G
 :  
undefined
 
Iron(II) sulphate heptahydrate ≥99% ACS
  0387-500G
 :  

 

  • Pack type:
    Bottle, amber
  • Pk:
    500 g
  • Seq:
    3645

 

 

  • Calcium:
    <0.005%
  • Chloride:
    ≤0.001%
  • Copper:
    ≤0.005%
  • Ferric Iron:
    ≤0.1%
  • Insolubles:
    ≤0.01%
  • Magnesium:
    ≤0.002%
  • Manganese:
    ≤0.05%
  • Phosphate:
    ≤0.001%
  • Potassium:
    ≤0.002%
  • Purity:
    ≥99%
  • Sodium:
    ≤0.02%
  • Zinc:
    ≤0.005%

 

 

 :
 :278.02 g/mol
 :13,04091
 :64 °C
 :1.9 g/cm³ (20 °C)
 :231-753-5
 :GHS07
 :Ambient
 :7782-63-0
 :FeSO₄·7H₂O
 :MFCD00149719