Iron(III) nitrate nonahydrate, Sigma-Aldrich®
Catalog # SIAL529303-25G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:404 g/mol
Merck Index:13,04057
Boiling Pt:125 °C (1013 hPa)
Melting Pt:47 °C
Density:1.68 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:233-899-5
UN:1466
ADR:5.1,III
:M-Clarity™ quality level = MQ100
:GHS03
:GHS07
CAS Number:7782-61-8
Formula:Fe(NO₃)₃·9H₂O
MDL Number:MFCD00149708