Order Entry
Austria
ContactUsLinkComponent
IHC reagent (mouse/rabbit-HRP), POLYVIEW®
IHC reagent (mouse/rabbit-HRP), POLYVIEW®
Catalog # ENZOADI9501130100
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
IHC reagent (mouse/rabbit-HRP), POLYVIEW®
Catalog # ENZOADI9501130100
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    100 Tests
  • Description:
    Anti-mouse/rabbit HRP detection reagent
  • Seq:
    02
  • Storage conditions:
    Long term
  • Storage temperature:
    4 °C
  • Application:
    IHC

Specifications

About this item

Immunohistochemistry (mouse/rabbit-HRP) detection reagent is a non-biotin one-step detection reagent.

  • Biotin-free nanopolymer detection circumvents endogenous biotin background
  • High intensity colour development, which delivers sharp and crisp staining
  • Used with blood smears, cytosmears, and cell preparations

Suitable for demonstrating antigens in formalin-fixed paraffin-embedded tissues and frozen sections.