Order Entry
Austria
ContactUsLinkComponent
IHC diluent (primary Ab)
IHC diluent (primary Ab)
Catalog # ENZOADI9502440250
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
IHC diluent (primary Ab)
Catalog # ENZOADI9502440250
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    250 mL
  • Description:
    IHC diluent (primary Ab)
  • Seq:
    02
  • Form:
    Liquid
  • Storage temperature:
    4 °C

Specifications

About this item

Immunohistochemistry diluent (primary Ab) can be used as a diluent to prepare primary antibodies and negative controls.

  • Optimal pH with stabilising proteins in the IHC diluent (primary Ab)
  • Maximum stability of primary antibodies
  • Good penetration of primary antibodies via selected concentrations of sodium chloride and detergents

Specifically designed to stabilise primary antibodies and reduce non-specific background staining observed during immunostaining.