

Scandium(III) nitrate hydate ≥99.99% (REO, rare earth oxide basis), REacton®
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:107552-14-7
Formula:Sc(NO₃)₃·xH₂O
MDL Number:MFCD00011220
:230.97 g/mol (anhy)
Merck Index:14,8392
Storage Temperature:Ambient
EINECS:236-701-5
UN:1477
ADR:5.1,III
:GHS03
:GHS07
Related Information
Frequently Bought Together