Order Entry
Austria
ContactUsLinkComponent
 
 
 
Copper(II) sulphate pentahydrate ≥98.5%, purified
 
undefined
 
 
Copper(II) sulphate pentahydrate ≥98.5%, purified
 

 

  • Assay:
    Min. 98.5 %
  • Cl (Chloride):
    Max. 20 ppm
  • Fe (Iron):
    Max. 100 ppm

 

Frequently Bought Together