IHC enzyme antigen retrieval reagent
Catalog # ENZOADI9502800015
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:15 mL
- Description:IHC enzyme antigen retrieval reagent
- Seq:02
Specifications
About this item
IHC enzyme antigen retrieval reagent is designed to unmask immunoreactive sites altered by fixation and/or the embedding process.
- Proteolytic enzyme solution that results in superior staining
- Reagent requires only a brief incubation at room temperature
- Allow to dilute the primary antibody 5 to 10 fold further and achieve optimal staining
Formulation: Liquid, enzyme antigen retrieval reagent
Appearance: Clear slightly viscous solution.