Order Entry
Austria
ContactUsLinkComponent
IHC mount, HIGHDEF®
IHC mount, HIGHDEF®
Catalog # ENZOADI9502610030
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
IHC mount, HIGHDEF®
Catalog # ENZOADI9502610030
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    30 mL
  • Description:
    IHC mount
  • Seq:
    02

Specifications

About this item

IHC mount is an aqueous-based permanent, high stability mounting medium designed for use with chromogenic labeling. Use for mounting of tissue specimens stained with peroxidase and alkaline phosphatase-based systems and various fluorescent dyes.

  • Flexible formulation optimised for AEC, DAB, Fast Red, BCIP/NBT, and BCIP/INT
  • High definition visualisation for IHC, IF, ISH, FISH
  • Allows permanent storage of tissue specimens without fading of the chromogens
  • No coverslip required
  • No exposure to organic fumes

Formulation: Liquid, mounting medium containing <0,1% sodium azide.