Order Entry
Austria
ContactUsLinkComponent
Copper(II) sulphate pentahydrate
Irritant
Environment
Copper(II) sulphate pentahydrate
Catalog # MOLE54865587
Supplier:  Molekula
CAS Number:  
Irritant
Environment
Copper(II) sulphate pentahydrate
Catalog # MOLE54865587
Supplier:  Molekula
CAS Number:  

Specifications

  • Pk:
    1 kg
  • Seq:
    4024

Specifications

About this item

 :
 :249.69 g/mol
Merck Index:13,02682
Density:2.284 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-847-6
UN:3077
ADR:9,III
REACH:01-2119520566-40
 :GHS07
 :GHS09
CAS Number:7758-99-8
Formula:CuSO₄·5H₂O
MDL Number:MFCD00149681