Order Entry
Austria
ContactUsLinkComponent
 
Copper(II) sulphate pentahydrate, EMSURE® ACS, ISO, Reag. Ph. Eur. for analysis, Supelco®
 :  Merck
 
Copper(II) sulphate pentahydrate, EMSURE® ACS, ISO, Reag. Ph. Eur. for analysis, Supelco®
 :  Merck
 
 
 

 

  • Assay (iodometric):
    99.0-100.5%
  • Insoluble matter:
    0.005% max
  • Chloride (Cl):
    0.0005% max
  • Total nitrogen (N):
    0.001% max
  • Calcium (Ca):
    0.005% max
  • Iron (Fe):
    0.003% max
  • Potassium (K):
    0.001% max
  • Sodium(Na):
    0.005% max
  • Nickel (Ni):
    0.005% max
  • Lead (Pb):
    0.005% max
  • Zinc (Zn):
    0.03% max

 

Frequently Bought Together