

Copper(II) sulphate pentahydrate USP, J.T.Baker®
Catalog # 1844-05
Supplier: Avantor
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:2,5 kg
- Seq:4076
Specifications
Specification Test Results
- Bulk Pharmaceutical Chemical:= 0.005 %
- USP - Calcium (Ca):≤0.005 %
- USP - Identification:Passes Test
- USP - Iron (Fe):≤0.003 %
- USP - Loss on Drying:33.0 - 36.5 %
- USP - Nickel (Ni):≤0.005 %
- USP - Potassium (K):≤0.01 %
- USP - Sodium (Na):≤0.02 %
- USP - Assay (CuSO₄) (dried basis):98.5 - 100.5 %
Specification Test Results
About this item
:
:249.69 g/mol
Merck Index:13,02682
Density:2.284 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-847-6
UN:3077
ADR:9,III
REACH:01-2119520566-40
:GHS07
:GHS09
CAS Number:7758-99-8
Formula:CuSO₄·5H₂O
MDL Number:MFCD00149681