


Copper(II) sulphate pentahydrate 99+% for analysis
:7758-99-8
:
:CuSO₄·5H₂O
:MFCD00149681
:249.69 g/mol
:13,02682
:110 °C (dec.)
:2,284 g/cm³ (20 °C)
:Ambient
:231-847-6
:3077
:9,III
:01-2119520566-40
:GHS05
:GHS07
:GHS09
- Appearance:Blue Crystalline powder or crystals
- Titration Iodometric:≥99.0 %
- Chloride (Cl):≤10 ppm
- Iron (Fe):≤50 ppm
- Lead (Pb):≤50 ppm
- Nickel (Ni):≤20 ppm
- Total nitrogen (as N):≤0.002 %
Frequently Bought Together