Order Entry
Austria
ContactUsLinkComponent
Copper(II) chloride dihydrate, EMSURE® ACS, Reag. Ph. Eur. for analysis, Supelco®
Corrosive
Irritant
Environment
Copper(II) chloride dihydrate, EMSURE® ACS, Reag. Ph. Eur. for analysis, Supelco®
Supplier:  Merck
Certificates
Corrosive
Irritant
Environment
Copper(II) chloride dihydrate, EMSURE® ACS, Reag. Ph. Eur. for analysis, Supelco®
Supplier:  Merck
Certificates

Specification Test Results

  • Solubility:
    1150 g/l (20 °C)
  • Melting Point:
    100 °C
  • Molar Mass:
    170.48 g/mol
  • Bulk Density:
    950 kg/m3
  • Density:
    2.54 g/cm3 (20 °C)
  • pH:
    3.0 - 3.8 (50 g/l, H2O, 20 °C)
  • Hazard Class:
    8
  • Hazard Class Description:
    Corrosive Material

Specification Test Results

Related Information

Frequently Bought Together