Modular heating blocks
Lieferant: OHAUS
Zertifikate
Über diesen Artikel
Modular blocks are constructed from a solid anodized aluminium block. The close contact of tubes-to-block walls allow for maximum heat retention. Each block has a thermometer well for measuring block temperature.
Achtung! Bitte beachten: To avoid possible electrical hazard, do not fill well or block with water or other fluids. Units are designed as a dry bath/incubator.
Frequently Bought Together