
Ammonium iron(II) sulphate hexahydrate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:7783-85-9
:
Formula:(NH₄)₂Fe(SO₄)₂·6H₂O
MDL Number:MFCD00150530
:392.14 g/mol
Melting Pt:100 °C
Density:1.864 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
EINECS:233-151-8
:GHS07
Storage Temperature:Ambient
Frequently Bought Together