Order Entry
Austria
ContactUsLinkComponent
Ammonium iron(II) sulphate hexahydrate ≥98.5%, TECHNICAL
Irritant
Ammonium iron(II) sulphate hexahydrate ≥98.5%, TECHNICAL
Supplier:  VWR Chemicals
Certificates
undefined
Irritant
Ammonium iron(II) sulphate hexahydrate ≥98.5%, TECHNICAL
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Assay:
    Min. 98.5 %

Specification Test Results

Frequently Bought Together