Order Entry
Austria
ContactUsLinkComponent
Ammonium iron(II) sulphate hexahydrate, EMSURE® for analysis, Supelco®
Irritant
Ammonium iron(II) sulphate hexahydrate, EMSURE® for analysis, Supelco®
Supplier:  Merck
Certificates
Irritant
Ammonium iron(II) sulphate hexahydrate, EMSURE® for analysis, Supelco®
Supplier:  Merck
Certificates

Specification Test Results

  • Assay (manganometric):
    99.0 - 101.5 %
  • pH-value (5 %; water):
    3.0 - 5.0
  • Chloride (Cl):
    ≤ 0.001 %
  • Phosphate (PO₄):
    ≤ 0.002 %
  • Iron(III)-salts (Fe³⁺):
    ≤ 0.02 %
  • Ca (Calcium):
    ≤ 0.002 %
  • Cu (Copper):
    ≤ 0.002 %
  • K (Potassium):
    ≤ 0.01 %
  • Mg (Magnesium):
    ≤ 0.01 %
  • Mn (Manganese):
    ≤ 0.05 %
  • Na (Sodium):
    ≤ 0.01 %
  • Pb (Lead):
    ≤ 0.001 %
  • Zn (Zinc):
    ≤ 0.003 %

Specification Test Results

Related Information

Certificates
Search by Lot Number
Frequently Bought Together