Order Entry
Austria
ContactUsLinkComponent
 
 
Ammonium iron(II) sulphate hexahydrate ≥99%
  A12473.36
 :  
 
Ammonium iron(II) sulphate hexahydrate ≥99%
  A12473.36
 :  

 

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g
  • Seq:
    3645

 

 

 :7783-85-9
 :
 :(NH₄)₂Fe(SO₄)₂·6H₂O
 :MFCD00150530
 :392.14 g/mol
 :14,521
 :∼ 100 °C (dec.)
 :1.864
 :233-382-4
 :GHS07
 :Ambient