ContactUsLinkComponent
Ammonium iron(II) sulphate hexahydrate 99+% for analysis
Irritant
Ammonium iron(II) sulphate hexahydrate 99+% for analysis
Certificates
undefined
Irritant
Ammonium iron(II) sulphate hexahydrate 99+% for analysis
Certificates
Specification Test Results
  • Appearance:
    Light green Crystalline powder or crystals
  • Titration with KMnO4:
    ≥99.0 %
  • Chloride (Cl):
    ≤10 ppm
  • Phosphate (PO4):
    ≤30 ppm
  • Copper (Cu):
    ≤20 ppm
  • Manganese (Mn):
    ≤500 ppm
  • Zinc (Zn):
    ≤30 ppm
  • Ferric iron (Fe 3+):
    ≤200 ppm (May increase due to inherent oxidation on storage)
Specification Test Results
Related Information
Certificates
Search by Lot Number
Frequently Bought Together