
Ammonium heptamolybdate tetrahydrate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:12054-85-2
:12027-67-7
Formula:(NH₄)₆Mo₇O₂₄·4H₂O
MDL Number:MFCD00167059
:1235.86 g/mol
Merck Index:13,00536
Density:2.498 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
EINECS:234-722-4
:GHS07
Storage Temperature:Ambient
Frequently Bought Together