Order Entry
Austria
ContactUsLinkComponent
 
Irritant
Ammonium heptamolybdate tetrahydrate, Supelco®
Catalog # 1.01181.1000
Supplier:  Merck
CAS Number:  
Irritant
Ammonium heptamolybdate tetrahydrate, Supelco®
Catalog # 1.01181.1000
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

Specification Test Results

  • Solubility:
    400 g/l (20 °C)
  • Melting Point:
    90 °C Elimination of water of crystallisation
  • Molar Mass:
    1235.86 g/mol
  • Bulk Density:
    800 kg/m3
  • Density:
    2.498 g/cm3 (20 °C)
  • pH:
    5.3 (50 g/l, H2O, 20 °C)

Specification Test Results

About this item

 :
 :1235.86 g/mol
Merck Index:13,00536
Density:2.498 g/cm³ (20 °C)
 :M-Clarity™ quality level = MQ100
EINECS:234-722-4
 :GHS07
Storage Temperature:Ambient
CAS Number:12054-85-2
 :12027-67-7
Formula:(NH₄)₆Mo₇O₂₄·4H₂O
MDL Number:MFCD00167059