Magnesium sulphate hydrate 99.0-101.0%, GPR RECTAPUR®
Catalog # 25169.295
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle for solids
- Pk:1 kg
- Seq:4024
- Environmentally Preferable:
Specifications
Specification Test Results
- Assay (on anhydrous substance):99.0 - 101.0%
- Heavy metals (as Pb):Max. 10 ppm
- Loss on ignition (450 ± 25 °C):22.0 - 28.0%
- Cl (Chloride):Max. 0.02%
- As (Arsenic):Max. 5 ppm
- Fe (Iron):Max. 10 ppm
Specification Test Results
About this item
:
:120.37 g/mol (anhy)
CAS Number:22189-08-8
:15244-36-7
Formula:MgSO₄·nH₂O
MDL Number:MFCD00011110
Density:2.45 g/cm³ (20 °C)