Order Entry
Austria
ContactUsLinkComponent
Magnesium sulphate heptahydrate ≥98%, purified
Magnesium sulphate heptahydrate ≥98%, purified
Supplier:  VWR Chemicals
Certificates
undefined
Magnesium sulphate heptahydrate ≥98%, purified
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Assay:
    Min. 98 %
  • Insoluble matter:
    Max. 0.05 %
  • Fe (Iron):
    Max. 20 ppm

Specification Test Results

Frequently Bought Together