Magnesium sulphate heptahydrate, EMSURE® ACS, Reag. Ph. Eur. for analysis, Supelco®
Supplier: Merck
Certificates
About this item
CAS Number:10034-99-8
:
Formula:MgSO₄·7H₂O
MDL Number:MFCD00149785
:246.48 g/mol
Merck Index:13,05715
Melting Pt:1124 °C
Density:2.66 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ300
Storage Temperature:Ambient
EINECS:231-298-2
Specification Test Results
- Solubility:710 g/l (20 °C)
- Molar Mass:246.48 g/mol
- Bulk Density:900 kg/m3
- Density:1.68 g/cm3 (20 °C)
- pH:5.0 - 8.0 (50 g/l, H2O, 25 °C)
Specification Test Results
Related Information
Frequently Bought Together