Order Entry
Austria
ContactUsLinkComponent
Magnesium sulphate heptahydrate USP, J.T.Baker®
Magnesium sulphate heptahydrate USP, J.T.Baker®
Catalog # 2505-07
Supplier:  Avantor
CAS Number:  
undefined
Magnesium sulphate heptahydrate USP, J.T.Baker®
Catalog # 2505-07
Supplier:  Avantor
CAS Number:  

Specifications

  • Pack type:
    PE pail, screw top
  • Pk:
    12 kg
  • Seq:
    4187

Specifications

Specification Test Results

  • Bulk Pharmaceutical Chemical:
    Passes Test
  • USP - Identification:
    Passes Test
  • USP - pH (1 in 20):
    5.0 - 9.2
  • USP - Loss on Ignition:
    40.0 - 52.0 %
  • USP - Chloride (Cl):
    ≤ 0.014 %
  • USP - Iron (Fe):
    ≤ 0.5 ppm
  • USP - Selenium (Se):
    ≤ 0.003 %
  • USP - Assay (MgSO₄) (anhydrous basis):
    99.0 - 100.5 %
  • USP - Endotoxin Concentration (2.5 EU/g max.):
    Passes Test

Specification Test Results

About this item

 :
 :246.48 g/mol
Merck Index:13,05715
Melting Pt:1124 °C
Density:2.66 g/cm³ (20 °C)
 :Management of Change (MOC) category = R
Storage Temperature:Ambient
EINECS:231-298-2
CAS Number:10034-99-8
Formula:MgSO₄·7H₂O
MDL Number:MFCD00149785