

Barium hydroxide octahydrate 98+% ACS
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:12230-71-6
:
Formula:Ba(OH)₂·8H₂O
MDL Number:MFCD00149152
:315.47 g/mol
Merck Index:14,977
Boiling Pt:780 °C (1013 hPa)
Melting Pt:78 °C
Density:2.18
Storage Temperature:Ambient
EINECS:241-234-5
UN:3262
ADR:8,III
:GHS05
:GHS07
Related Information
Frequently Bought Together