Order Entry
Austria
ContactUsLinkComponent
Magnesium perchlorate hexahydrate, Supelco®
Magnesium perchlorate hexahydrate, Supelco®
Catalog # 63043-50ML-F
Supplier:  Merck
CAS Number:  
Magnesium perchlorate hexahydrate, Supelco®
Catalog # 63043-50ML-F
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    50 mL
  • Seq:
    2370
  • Environmentally Preferable:

Specifications

About this item

 :
 :331.3 g/mol
EINECS:233-108-3
UN:1475
CAS Number:13446-19-0
Formula:Cl₂MgO₈·6H₂O
MDL Number:MFCD00149778
Melting Pt:185…190 °C
Density:1.98 g/cm³ (20 °C)
ADR:5.1 II
 :M-Clarity™ quality level = MQ100