Magnesium chloride hexahydrate, crystallised, EMPROVE® API Ph. Eur., BP, JPC, USP suitable for use as an active pharmaceutical ingredient, SAFC®
:7791-18-6
:
:MgCl₂·6H₂O
:MFCD00149781
:203.3 g/mol
:13,05684
:1412 °C (1013 hPa)
:117 °C
:1.57 g/cm³ (25 °C)
:Ambient
:232-094-6
- Assay (complexometric):99.0 - 101.0 %
- Identity:passes test
- Appearance of solution:passes test
- Insoluble matter:≤ 0.005 %
- Acidity or alkalinity:passes test
- pH-value (5 %; water):5.0 - 7.0
- Bromide (Br):≤ 0.05 %
- Sulfate (SO₄):≤ 0.005 %
- Heavy metals (as Pb):≤ 0.001 %
- Al (Aluminium):≤ 0.0001 %
- As (Arsenic):≤ 0.0002 %
- Ba (Barium):passes test
- Ca (Calcium):≤ 0.01 %
- Fe (Iron):≤ 0.0005 %
- Hg (Mercury):≤ 0.0001 %
- K (Potassium):≤ 0.0500 %
- NH₄ (Ammonium):≤ 0.005 %
- Pb (Lead):≤ 0.0004 %
- Water:51.0 - 55.0 %
- Total aerobic microbial count:≤ 100 CFU/g
- Total yeast and mould count:≤ 100 CFU/g
- Endotoxins:≤ 3.0 I.U./g
- Residual solvents (Ph Eur/USP/ICH):excluded by manufacturing process
Frequently Bought Together