
Magnesium bromide hexahydrate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:13446-53-2
:
Formula:Br₂Mg·6H₂O
MDL Number:MFCD00149780
:292.2 g/mol
Melting Pt:146 °C
Density:2 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
EINECS:232-170-9
:GHS07
Frequently Bought Together