
Magnesium bromide hexahydrate ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:13446-53-2
:
Formula:Br₂Mg·6H₂O
MDL Number:MFCD00149780
:292.2 g/mol
Melting Pt:172 °C
Density:2.00
EINECS:232-170-9
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together