Sodium tungstate dihydrate ≥95%
About this item
Purity: 95% CAS No.: 10213-10-2 Molecular Formula: Na2WO4.2H2O Formula Weight: 329.86 (293.83anhy) Physical Form: Crystalline Density: 3.250 Appearance: White Odour: Odourless Melting Point: 100° -2H2O Solubility: Soluble in water. Insoluble in alcohol Merck Reference: 14,8698 MDL No.: MFCD00149190
- CAS Number:10213-10-2
- Formula:Na₂WO₄·2H₂O
- MDL Number:MFCD00149190
- Molecular Weight:329.85 g/mol
- Merck Index:14,8698
- Melting Pt:100 °C (-2H₂O)
- Density:3.250
- Storage Temperature:Ambient
- EINECS:236-743-4
2 Options Available Below
Product Details & Documents
Purity: 95% CAS No.: 10213-10-2 Molecular Formula: Na2WO4.2H2O Formula Weight: 329.86 (293.83anhy) Physical Form: Crystalline Density: 3.250 Appearance: White Odour: Odourless Melting Point: 100° -2H2O Solubility: Soluble in water. Insoluble in alcohol Merck Reference: 14,8698 MDL No.: MFCD00149190
- CAS Number:10213-10-2
- Formula:Na₂WO₄·2H₂O
- MDL Number:MFCD00149190
- Molecular Weight:329.85 g/mol
- Merck Index:14,8698
- Melting Pt:100 °C (-2H₂O)
- Density:3.250
- Storage Temperature:Ambient
- EINECS:236-743-4
Documents
Product Family Options
Product Information
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



